Learn More
Felix Storch Inc ACCUCOLD ACCUCOLD ARG12ML Performance Medical-Laboratory Refrigerator 12 Cu. Ft. with Solid Door
SDP
Catalog No. 501959436
Encompass_Preferred
ACCUCOLD ARS12ML Performance Medical-Laboratory Refrigerator 12 Cu. Ft. with Solid Door with Advanced Temperature Control, cycle defrost and forced air cooling. Adjustable operating control range of 2C to 10C with Intelligent Microprocessor Digital Controller designed to provide +/- 1C variation. Multiple Audio and Visual Alarms for Safer Temperature Monitoring. Digital display of the min/max temperature (updated in 15 minute increments). Remote alarm contacts. LED lighting, adjustable shelves, 1/2' probe hole, reversible door. Meets EPA and SNAP requirements. Eco-Friendly using hydrocarbons for a greener global footprint. Compatible with Omnicell, AcuDose, and Pyxis (not included), ADA Option available. 2 Year Warranty Parts and Labor, 5-year compressor: https://www.accucold.com/product/ARS12ML
Catalog No. 50-195-9436 Supplier Felix Storch Inc Supplier No. ARS12ML
Add to Cart
Add to Cart
Request a Quote
ACCUCOLD ARS12ML Performance Medical-Laboratory Refrigerator 12 Cu. Ft. with Solid Door with Advanced Temperature Control, cycle defrost and forced air cooling. Adjustable operating control range of 2C to 10C with Intelligent Microprocessor Digital Controller designed to provide +/- 1C variation. Multiple Audio and Visual Alarms for Safer Temperature Monitoring. Digital display of the min/max temperature (updated in 15 minute increments). Remote alarm contacts. LED lighting, adjustable shelves, 1/2' probe hole, reversible door. Meets EPA and SNAP requirements. Eco-Friendly using hydrocarbons for a greener global footprint. Compatible with Omnicell, AcuDose, and Pyxis (not included), ADA Option available. 2 Year Warranty Parts and Labor, 5-year compressor: https://www.accucold.com/product/ARS12ML