Learn More
Felix Storch Inc ACCUCOLD ACCUCOLD ARG1ML Performance Medical-Laboratory Refrigerator 1 Cu. Ft. with Glass Door
SDP
Catalog No. 501959430
Encompass_Preferred
ACCUCOLD ARG1ML Performance Medical-Laboratory Refrigerator 1 Cu. Ft. with Glass Door with Advanced Temperature Control, cycle defrost and forced air cooling. Adjustable operating control range of 2C to 10C with Intelligent Microprocessor Digital Controller designed to provide +/- 1C variation. Multiple Audio and Visual Alarms for Safer Temperature Monitoring. Digital display of the min/max temperature (updated in 15 minute increments). Remote alarm contacts. LED lighting, adjustable shelves, 1/2' probe hole, reversible and self closing door. Meets EPA and SNAP requirements. Eco-Friendly using hydrocarbons for a greener global footprint. Compatible with Omnicell, AcuDose, and Pyxis (not included), ADA Option available. 2 Year Warranty Parts and Labor, 5-year compressor: https://www.accucold.com/product/ARG1ML
Catalog No. 50-195-9430 Supplier Felix Storch Inc Supplier No. ARG1ML
ACCUCOLD ARG1ML Performance Medical-Laboratory Refrigerator 1 Cu. Ft. with Glass Door with Advanced Temperature Control, cycle defrost and forced air cooling. Adjustable operating control range of 2C to 10C with Intelligent Microprocessor Digital Controller designed to provide +/- 1C variation. Multiple Audio and Visual Alarms for Safer Temperature Monitoring. Digital display of the min/max temperature (updated in 15 minute increments). Remote alarm contacts. LED lighting, adjustable shelves, 1/2' probe hole, reversible and self closing door. Meets EPA and SNAP requirements. Eco-Friendly using hydrocarbons for a greener global footprint. Compatible with Omnicell, AcuDose, and Pyxis (not included), ADA Option available. 2 Year Warranty Parts and Labor, 5-year compressor: https://www.accucold.com/product/ARG1ML