Learn More
Felix Storch Inc ACCUCOLD ACCUCOLD ARG23ML Performance Medical-Laboratory Refrigerator 23 Cu. Ft. with Glass Door and casters
SDP
Catalog No. 501959431
Encompass_Preferred
ACCUCOLD ARG23ML Performance Medical-Laboratory Refrigerator 23 Cu. Ft. with Glass Door with Advanced Temperature Control, cycle defrost and forced air cooling. Adjustable operating control range of 2C to 10C with Intelligent Microprocessor Digital Controller designed to provide +/- 1C variation. Multiple Audio and Visual Alarms for Safer Temperature Monitoring. Digital display of the min/max temperature (updated in 15 minute increments). LED lighting, adjustable shelves, and self closing door. Meets EPA and SNAP requirements. Eco-Friendly using hydrocarbons for a greener global footprint. 2 Year Warranty Parts and Labor, 5-year compressor: https://www.accucold.com/product/ARG23ML
Catalog No. 50-195-9431 Supplier Felix Storch Inc Supplier No. ARG23ML
Add to Cart
Add to Cart
Request a Quote
ACCUCOLD ARG23ML Performance Medical-Laboratory Refrigerator 23 Cu. Ft. with Glass Door with Advanced Temperature Control, cycle defrost and forced air cooling. Adjustable operating control range of 2C to 10C with Intelligent Microprocessor Digital Controller designed to provide +/- 1C variation. Multiple Audio and Visual Alarms for Safer Temperature Monitoring. Digital display of the min/max temperature (updated in 15 minute increments). LED lighting, adjustable shelves, and self closing door. Meets EPA and SNAP requirements. Eco-Friendly using hydrocarbons for a greener global footprint. 2 Year Warranty Parts and Labor, 5-year compressor: https://www.accucold.com/product/ARG23ML