Learn More
Felix Storch Inc ACCUCOLD ACCUCOLD ARS23ML Performance Medical-Laboratory Refrigerator 23 Cu. Ft. with Solid Door and casters
SDP
Catalog No. 501959439
Encompass_Preferred
ACCUCOLD ARS23ML Performance Medical-Laboratory Refrigerator 23 Cu. Ft. with Solid Door with Advanced Temperature Control, cycle defrost and forced air cooling. Adjustable operating control range of 2C to 10C with Intelligent Microprocessor Digital Controller designed to provide +/- 1C variation. Multiple Audio and Visual Alarms for Safer Temperature Monitoring. Digital display of the min/max temperature (updated in 15 minute increments). LED lighting, adjustable shelves, and self closing door. Meets EPA and SNAP requirements. Eco-Friendly using hydrocarbons for a greener global footprint. ADA Option available. 2 Year Warranty Parts and Labor, 5-year compressor: https://www.accucold.com/product/ARS23ML
Catalog No. 50-195-9439 Supplier Felix Storch Inc Supplier No. ARS23ML
Add to Cart
Add to Cart
Request a Quote
ACCUCOLD ARS23ML Performance Medical-Laboratory Refrigerator 23 Cu. Ft. with Solid Door with Advanced Temperature Control, cycle defrost and forced air cooling. Adjustable operating control range of 2C to 10C with Intelligent Microprocessor Digital Controller designed to provide +/- 1C variation. Multiple Audio and Visual Alarms for Safer Temperature Monitoring. Digital display of the min/max temperature (updated in 15 minute increments). LED lighting, adjustable shelves, and self closing door. Meets EPA and SNAP requirements. Eco-Friendly using hydrocarbons for a greener global footprint. ADA Option available. 2 Year Warranty Parts and Labor, 5-year compressor: https://www.accucold.com/product/ARS23ML