Learn More
Felix Storch Inc ACCUCOLD ACCUCOLD ARS29ML Performance Medical-Laboratory Refrigerator 49 Cu. Ft. with 2 Solid Door and casters
SDP
Catalog No. 501959441
Encompass_Preferred
ACCUCOLD ARS49ML Performance Medical-Laboratory Refrigerator 49 Cu. Ft. with 2 Solid Door with Advanced Temperature Control, cycle defrost and forced air cooling. Adjustable operating control range of 2C to 10C with Intelligent Microprocessor Digital Controller designed to provide +/- 1C variation. Multiple Audio and Visual Alarms for Safer Temperature Monitoring. Digital display of the min/max temperature (updated in 15 minute increments). LED lighting, adjustable shelves, self closing door. Meets EPA and SNAP requirements. Eco-Friendly using hydrocarbons for a greener global footprint. 2 Year Warranty Parts and Labor, 5-year compressor: https://www.accucold.com/product/ARS49ML
Catalog No. 50-195-9441 Supplier Felix Storch Inc Supplier No. ARS49ML
Add to Cart
Add to Cart
Request a Quote
ACCUCOLD ARS49ML Performance Medical-Laboratory Refrigerator 49 Cu. Ft. with 2 Solid Door with Advanced Temperature Control, cycle defrost and forced air cooling. Adjustable operating control range of 2C to 10C with Intelligent Microprocessor Digital Controller designed to provide +/- 1C variation. Multiple Audio and Visual Alarms for Safer Temperature Monitoring. Digital display of the min/max temperature (updated in 15 minute increments). LED lighting, adjustable shelves, self closing door. Meets EPA and SNAP requirements. Eco-Friendly using hydrocarbons for a greener global footprint. 2 Year Warranty Parts and Labor, 5-year compressor: https://www.accucold.com/product/ARS49ML