Learn More
Cell Systems Corporation Medium Formulated at Normal Blood Glucose Level Without Serum and With CultureBoost (4N3-500)
SDP
Catalog No. NC1876704
Encompass_Preferred
Our popular medium designed with normal blood glucose levels and without serum can be tailored to specific cell culture needs. For complete medium, add 5-20% serum (defined FBS or equivalent) (not included) and the included CultureBoost supplement. This kit consists of 500mL Normal Glucose Medium, 5mL CultureBoost (with animal derived growth factors) and 10mL Attachment Factor (extracellular matrix reagent for coating flasks). 4N3-500 is certified and intended for experimental application. The media is phenol-red free with no added hormones or antibiotics for optimized experimental conditions. It is certified to be used with more than 100 human and animal cell cultures and cell lines. Large quantities? Contact Fisher Scientific sales rep to discuss your bulk order needs. Tight deadline? Our products are available for next-day delivery after the purchase order is received. Maximize your results! Try our Primary Human Cells and Passage Reagent (4Z0-800), both available for purchase.
Catalog No. NC1876704 Supplier Cell Systems Corporation Supplier No. 4N3500
Our popular medium designed with normal blood glucose levels and without serum can be tailored to specific cell culture needs. For complete medium, add 5-20% serum (defined FBS or equivalent) (not included) and the included CultureBoost supplement. This kit consists of 500mL Normal Glucose Medium, 5mL CultureBoost (with animal derived growth factors) and 10mL Attachment Factor (extracellular matrix reagent for coating flasks). 4N3-500 is certified and intended for experimental application. The media is phenol-red free with no added hormones or antibiotics for optimized experimental conditions. It is certified to be used with more than 100 human and animal cell cultures and cell lines. Large quantities? Contact Fisher Scientific sales rep to discuss your bulk order needs. Tight deadline? Our products are available for next-day delivery after the purchase order is received. Maximize your results! Try our Primary Human Cells and Passage Reagent (4Z0-800), both available for purchase.