Learn More
Electron Microscopy Sciences Medium SEM Clamp, 25 x 15 mm, 3.2 mm Pin diameter
SDP
Catalog No. 501927144
Encompass_Preferred
Scanning electron microscope clamp for samples up to 7 mm overall thickness including small tubes, strips, wires, and smaller flat samples. Dimensions: 23 x 20 x 9 mm. Perfect for small tubes, strips, wires, and smaller flat samples. Overall Dimensions: 33 x 25 x 9 mm (1.3" x 1" x 0.35"). Made from machined aluminum with brass screws.
Size: 25 x 15 mm
Pin diameter: 3.2 mm
Catalog No. 50-192-7144 Supplier Electron Microscopy Sciences Supplier No. 7595403
Scanning electron microscope clamp for samples up to 7 mm overall thickness including small tubes, strips, wires, and smaller flat samples. Dimensions: 23 x 20 x 9 mm. Perfect for small tubes, strips, wires, and smaller flat samples. Overall Dimensions: 33 x 25 x 9 mm (1.3" x 1" x 0.35"). Made from machined aluminum with brass screws.
Size: 25 x 15 mm
Pin diameter: 3.2 mm