Learn More
DR Instruments Med-Student Sphygmomanometer Kit
SDPSCI_ED
Catalog No. S20676
Change view
Click to view available options
Size:
Standard
Catalog No. Size
S20676 Standard
1 options
California Proposition 65 WARNING: Cancer and Reproductive Harm
This item is intended for educational purposes only.
Catalog No. S20676 Supplier DR Instruments Supplier No. DR109747
Add to Cart
Add to Cart
California Proposition 65 WARNING: Cancer and Reproductive Harm
This item is intended for educational purposes only.
The Med-Student Sphygmomanometer Kit contains professional quality cuff and MDF™ dual-head stethoscope.
Kit contains:
  • Aneroid Sphygmomanometer - Standard size cuff made of nylon with index and range markings for proper positioning. Black inflation bulb and chrome plated inflation valve with traditional thumb-screw air control. Equipped with no-pin stop manometer gauge graduated to 300 mmHg.
  • MDF Dual Head Stethoscope - Bowles and deep cone-shaped Bell with a full-rotation acoustic valve stem and Headset Spring.
  • Black zippered carrying case
Specifications
Display Analog
Size Standard
For Use With (Equipment) Blood Pressure Cuff
For Use With (Application) For medical students and professionals
Includes Aneroid Sphygmomanometer, MDF Dual Head Stethoscope, Black Zippered Carrying Case
Material Nylon
Product Type Sphygmomanometer Kit
Pressure 300 mmHg