Learn More
McKesson General Medical Rolyan Synergy Thermoplastic Splinting Material, Maximum Stretch Resistance, Low Drape, 1/8 in
Catalog No. NC3311818
Encompass_Preferred
Rolyan Synergy Thermoplastic Splinting Material is a versatile, polycaprolactone-based sheet designed for the fabrication of medical splints and orthoses. Its unique properties, including maximum stretch resistance and minimal conformability, provide clinicians with exceptional control during shaping—preventing unwanted deformation. The smooth, uncoated surface ensures permanent self-bonding when heated, enabling easy adjustments and reuse. This non-toxic, latex-free material is highly beneficial for a wide range of therapeutic applications.
Catalog No. NC3311818 Supplier McKesson General Medical Supplier No. 7355PK
Rolyan Synergy Thermoplastic Splinting Material is a versatile, polycaprolactone-based sheet designed for the fabrication of medical splints and orthoses. Its unique properties, including maximum stretch resistance and minimal conformability, provide clinicians with exceptional control during shaping—preventing unwanted deformation. The smooth, uncoated surface ensures permanent self-bonding when heated, enabling easy adjustments and reuse. This non-toxic, latex-free material is highly beneficial for a wide range of therapeutic applications.