Learn More
Medicus Health Med Vial Holder, Clamp Mount
SDP
Catalog No. NC2199862
Encompass
∙ Clamp Mount Vial Holder allows safe, hands-free access to med vials ∙ Dimensions: 6.25""""W x 4.5""""D x 5.25""""H ∙ Allows for safe removal of vial contents reducing risk of accidental sticks ∙ Easy viewing of labels helps prevent medication errors ∙ Downward vial position assures complete usage of expensive medication ∙ Clamps to any table surface or stand up to 1.25″ thick ∙ Made of high grade anodized aluminum
This item is not returnable. View return policy
Catalog No. NC2199862 Supplier Medicus Health Supplier No. 2942M2
∙ Clamp Mount Vial Holder allows safe, hands-free access to med vials ∙ Dimensions: 6.25""""W x 4.5""""D x 5.25""""H ∙ Allows for safe removal of vial contents reducing risk of accidental sticks ∙ Easy viewing of labels helps prevent medication errors ∙ Downward vial position assures complete usage of expensive medication ∙ Clamps to any table surface or stand up to 1.25″ thick ∙ Made of high grade anodized aluminum