Learn More
Aviva Systems Biology Corp MED14 Antibody - middle region (ARP58139_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58139_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MED14. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human MED14. Alternate names: CRSP150, CRSP2, CSRP, CXorf4, DRIP150, EXLM1, MGC104513, RGR1, TRAP170ApplicationWB, CHIP. Species reactivity: Human, Mouse, Rabbit
Catalog No. 50-286-443