Learn More
Aviva Systems Biology Corp MED16 Antibody - N-terminal region (ARP31452_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31452_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MED16. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MED16. Alternate names: DRIP92, THRAP5, TRAP95ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-52828