Learn More
Aviva Systems Biology Corp MED18 Antibody - middle region (ARP62133_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62133_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MED18. It was validated on Western Blot. . Alternate names: FLJ20045, p28bApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-582