Learn More
ABclonal Technology MED18 Rabbit pAb
SDP
Supplier:  ABclonal Technology A145881000UL
Encompass_Preferred
MED18 is a component of the Mediator complex, which is a coactivator for DNA-binding factors that activate transcription via RNA polymerase II (Sato et al., 2003 [PubMed 12584197]).
Catalog No. 50-001-07072