Learn More
Aviva Systems Biology Corp MED21 Antibody - C-terminal region (ARP35727_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35727_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MED21. It was validated on Western Blot. . Alternate names: SRB7, SURB7ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-284-471