Learn More
Aviva Systems Biology Corp MED23 Antibody - N-terminal region (ARP34880_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34880_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MED23. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MED23. Alternate names: CRSP130, CRSP133, CRSP3, DKFZp434H0117, DRIP130, SUR2, MRT18, SUR-2, ARC130ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-55598