Learn More
Aviva Systems Biology Corp MED25 Antibody - N-terminal region (ARP50698_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50698_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MED25. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human MED25. Alternate names: ACID1, ARC92, CMT2B2, DKFZp434K0512, MGC70671, P78, PTOV2, TCBAP0758ApplicationWB, CHIP. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rat, Zebrafish
Catalog No. 50-285-528