Learn More
Aviva Systems Biology Corp MED26 Antibody - N-terminal region (ARP33745_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33745_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MED26. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MED26. Alternate names: CRSP7, CRSP70ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55132