Learn More
Aviva Systems Biology Corp MED27 Antibody - middle region (ARP33743_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33743_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MED27. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MED27. Alternate names: CRAP34, CRSP34, CRSP8, MGC11274, TRAP37ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55131