Learn More
Aviva Systems Biology Corp MED4 Antibody - C-terminal region (ARP50562_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50562_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MED4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human MED4. Alternate names: DRIP36, FLJ10956, HSPC126, RP11-90M2.2, TRAP36, VDRIP, ARC36ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53635