Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProSci MED4 Peptide
SDP

Catalog No. 16919805
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-919-805 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-919-805 Supplier ProSci Supplier No. 4729P0050
Only null left
Add to Cart
Add to Cart

Used for Blocking Activity

MED4 peptide is used for blocking the activity of MED4 antibody.

Specifications

Gene ID (Entrez) 29079
Species Human
Accession Number NP_054885
Common Name mediator complex subunit 4
Concentration 200 μg/mL
Content And Storage Store MED4 peptide at -20°C, stable for one year.
Format Liquid
Formulation PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide
For Use With (Application) Blocking Assay
Gene Symbol MED4
Quantity 0.05 mg
Sequence 17 amino acids near the carboxy terminus of human MED4.
Synonym ARC36, VDRIP, DRIP36, TRAP36, HSPC126, ARC36, Mediator of RNA polymerase II transcription subunit 4, Activator-recruited cofactor 36 kDa component
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.