Learn More
Aviva Systems Biology Corp MED8 Antibody - N-terminal region (ARP52440_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52440_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MED8. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MED8. Alternate names: ARC32, MGC17544, MGC19641ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60743