Description
- Monomeric formulation which upon contact with an alkaline pH environment, polymerizes to form a thin flexible waterproof bandage.
- Polymerizes in apporximately one second after contact with moist wound surfaces.
- Polymerization is slower on a dry field.
- Also seals off exposed nerve endings.
Specifications
Specifications
| For Use With (Application) | Neuroscience |
| Size | 2 mL |
| Includes | Pack of 5 |