Learn More
Stoelting™ Medbond Tissue Glue
SDP
Catalog No. 10000648 Shop All Stoelting Co Products
Change view
Click to view available options
Includes:
Box of 25
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
10-000-648 Box of 25
10-000-647 Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-000-648 Supplier Stoelting™ Supplier No. 50479
Add to Cart
Add to Cart
Stoelting™ Medbond is a hemostatic agents that seals off capillary ends.
  • Monomeric formulation which upon contact with an alkaline pH environment, polymerizes to form a thin flexible waterproof bandage.
  • Polymerizes in apporximately one second after contact with moist wound surfaces.
  • Polymerization is slower on a dry field.
  • Also seals off exposed nerve endings.
Specifications
For Use With (Application) Neuroscience
Size 2 mL
Includes Box of 25