Learn More
Medicus Health Refrigerator MedBox with Programmable Electronic Lock, 10.9"W x 16.5"D x 6"H
SDP
Catalog No. NC2045470
Encompass
∙ Programmable Medication Box with Electronic Lock allows you to program your own combination and eliminates the need for keys ∙ Medication Box with easy to use eco-friendly electronic programmable lock with a battery life of over 10 years! ∙ Box Dimensions: 10.9"W x 16.5"D x 6"H∙ Lock is easy to program and comes with three different passwords: master, service and user∙ All programming is controlled via the master password with no expensive key to lose or become defective∙ Lock meets IP55 rating for humidity and moisture so it's perfect for mounting inside refrigerators∙ Helps comply with Joint Commission regulations to keep high risk medications locked up at all times∙ High density polyethylene box resists most solvents & chemicals∙ All boxes complete mounting kit for wire and glass shelf refrigerators
Catalog No. NC2045470 Supplier Medicus Health Supplier No. 2748M3BLUE
∙ Programmable Medication Box with Electronic Lock allows you to program your own combination and eliminates the need for keys ∙ Medication Box with easy to use eco-friendly electronic programmable lock with a battery life of over 10 years! ∙ Box Dimensions: 10.9"W x 16.5"D x 6"H∙ Lock is easy to program and comes with three different passwords: master, service and user∙ All programming is controlled via the master password with no expensive key to lose or become defective∙ Lock meets IP55 rating for humidity and moisture so it's perfect for mounting inside refrigerators∙ Helps comply with Joint Commission regulations to keep high risk medications locked up at all times∙ High density polyethylene box resists most solvents & chemicals∙ All boxes complete mounting kit for wire and glass shelf refrigerators