Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC MedChemExpress Antibody, B7-H3 Target, Human, Anti-tumor Activity
SDP

Catalog No. NC3975415
Encompass_Preferred

This antibody targets B7-H3 and exhibits anti-tumor activity. It can be utilized for the synthesis of antibody-drug conjugates (ADCs) and is suitable for research in areas such as small cell lung cancer, relapsed or refractory osteosarcoma, and advanced solid tumors.

  • Target: B7-H3
  • Exhibits anti-tumor activity
  • Can be studied for small cell lung cancer, relapsed or refractory osteosarcoma, and advanced solid tumors
  • Human IgG1 kappa isotype
  • Human species reactivity
  • Applications include ELISA, FACS, and functional assay
  • Unconjugated format
  • Can be reconstituted/diluted with sterile PBS or saline
  • Immobilized huB7H3-His can bind Risvutatug with an EC50 of 13.41 ng/mL
  • Purity: 99.57%
  • Molecular weight: 143.94 kDa
  • Appearance: liquid
  • Color: colorless to light yellow

Catalog No. NC3975415 Supplier Medchemexpress LLC Supplier No. HYP99153710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This antibody targets B7-H3 and exhibits anti-tumor activity. It can be utilized for the synthesis of antibody-drug conjugates (ADCs) and is suitable for research in areas such as small cell lung cancer, relapsed or refractory osteosarcoma, and advanced solid tumors.

  • Target: B7-H3
  • Exhibits anti-tumor activity
  • Can be studied for small cell lung cancer, relapsed or refractory osteosarcoma, and advanced solid tumors
  • Human IgG1 kappa isotype
  • Human species reactivity
  • Applications include ELISA, FACS, and functional assay
  • Unconjugated format
  • Can be reconstituted/diluted with sterile PBS or saline
  • Immobilized huB7H3-His can bind Risvutatug with an EC50 of 13.41 ng/mL
  • Purity: 99.57%
  • Molecular weight: 143.94 kDa
  • Appearance: liquid
  • Color: colorless to light yellow

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.