Learn More
Medchemexpress LLC MedChemExpress ITK PROTAC Degrader, ITK Target, 1022.20 MW, 98.31% Purity
SDP
Catalog No. NC3981373
Encompass
BSJ-05-037 is an ITK PROTAC degrader that effectively targets and degrades ITK in T-cell lymphoma cell lines. It blocks the activation of the NF-κB/GATA-3 signaling pathway, inhibits PLCγ1 phosphorylation, and reduces the proliferative capacity of T-cell lymphoma cells. This compound enhances the sensitivity of T-cell lymphoma cells to chemotherapy and is applicable to research related to T-cell lymphoma.
  • Targets and degrades ITK in T-cell lymphoma cell lines.
  • Blocks the activation of the NF-κB/GATA-3 signaling pathway.
  • Inhibits PLCγ1 phosphorylation.
  • Reduces the proliferative capacity of T-cell lymphoma cells.
  • Enhances the sensitivity of T-cell lymphoma cells to chemotherapy.
  • Decreases tumor volume in mouse models.
  • Reverses chemotherapy resistance in mouse models.
Catalog No. NC3981373 Supplier Medchemexpress LLC Supplier No. HY18520610MG
BSJ-05-037 is an ITK PROTAC degrader that effectively targets and degrades ITK in T-cell lymphoma cell lines. It blocks the activation of the NF-κB/GATA-3 signaling pathway, inhibits PLCγ1 phosphorylation, and reduces the proliferative capacity of T-cell lymphoma cells. This compound enhances the sensitivity of T-cell lymphoma cells to chemotherapy and is applicable to research related to T-cell lymphoma.
  • Targets and degrades ITK in T-cell lymphoma cell lines.
  • Blocks the activation of the NF-κB/GATA-3 signaling pathway.
  • Inhibits PLCγ1 phosphorylation.
  • Reduces the proliferative capacity of T-cell lymphoma cells.
  • Enhances the sensitivity of T-cell lymphoma cells to chemotherapy.
  • Decreases tumor volume in mouse models.
  • Reverses chemotherapy resistance in mouse models.