Learn More
Medchemexpress LLC MEDCHEMEXPRESS LLC HMG-CoA Reductase Inhibitor, Atorvastatin, 10 mg, 99.60% Purity
SDP
Catalog No. NC3984495
Encompass_Preferred
Atorvastatin is an orally active HMG-CoA reductase inhibitor that effectively decreases blood lipids. It inhibits human SV-SMC proliferation and invasion. This product is suitable for various research applications.
  • Orally active HMG-CoA reductase inhibitor.
  • Effectively decreases blood lipids.
  • Inhibits human SV-SMC proliferation and invasion with IC50s of 0.39 μM and 2.39 μM.
  • For research purposes only.
  • High purity of 99.60%.
Catalog No. NC3984495 Supplier Medchemexpress LLC Supplier No. HYB058910MG
Atorvastatin is an orally active HMG-CoA reductase inhibitor that effectively decreases blood lipids. It inhibits human SV-SMC proliferation and invasion. This product is suitable for various research applications.
  • Orally active HMG-CoA reductase inhibitor.
  • Effectively decreases blood lipids.
  • Inhibits human SV-SMC proliferation and invasion with IC50s of 0.39 μM and 2.39 μM.
  • For research purposes only.
  • High purity of 99.60%.