Learn More
Medchemexpress LLC Medetomidine (hydrochloride) | 86347-15-1 | 99.8% | 236.74 | 1 ML
SDP
Catalog No. 5000384521
Encompass_Preferred
Medetomidine hydrochloride is an orally active α2-adrenoceptor agonist with sedative and analgesic effects. It can cause peripheral vasoconstriction through the activation of α2 adrenoceptors on blood vessels. It is for research use only.
  • Orally active α2-adrenoceptor agonist (Ki: 1.08 nM)
  • Sedative and analgesic effects
  • Causes peripheral vasoconstriction through activation of α2 adrenoceptors
  • Inhibits aldosterone release from adrenocortical cell suspension in vitro
  • Activates a kicking response in Cyprids in vitro
  • Increases cellular cAMP production by activating β-like receptors in CHO cells in vitro
  • Induces sedation in cats in vivo
  • Shows sedative and analgesic effects in dogs in vivo
  • Protects against Diazinon-induced toxicosis in mice in vivo
Catalog No. 50-003-84521 Supplier Medchemexpress LLC Supplier No. HY17034B10MM/1ML
Medetomidine hydrochloride is an orally active α2-adrenoceptor agonist with sedative and analgesic effects. It can cause peripheral vasoconstriction through the activation of α2 adrenoceptors on blood vessels. It is for research use only.
  • Orally active α2-adrenoceptor agonist (Ki: 1.08 nM)
  • Sedative and analgesic effects
  • Causes peripheral vasoconstriction through activation of α2 adrenoceptors
  • Inhibits aldosterone release from adrenocortical cell suspension in vitro
  • Activates a kicking response in Cyprids in vitro
  • Increases cellular cAMP production by activating β-like receptors in CHO cells in vitro
  • Induces sedation in cats in vivo
  • Shows sedative and analgesic effects in dogs in vivo
  • Protects against Diazinon-induced toxicosis in mice in vivo