Learn More
Medchemexpress LLC Medetomidine (hydrochloride) | 86347-15-1 | 99.8% | 236.74 | 100 MG
SDP
Catalog No. 5000385727
Encompass_Preferred
Medetomidine hydrochloride is an analytical standard of Medetomidine (hydrochloride), intended for research and analytical applications. This orally active α2-adrenoceptor agonist, with a Ki of 1.08 nM, exhibits sedative and analgesic effects, and can induce peripheral vasoconstriction. It serves as a reference standard for assays and is commonly utilized in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Analytical standard for research and analytical applications
  • Orally active α2-adrenoceptor agonist (Ki: 1.08 nM)
  • Provides sedative and analgesic effects
  • Induces peripheral vasoconstriction
  • Reference standard for assays
  • Suitable for HPLC, GC, and MS research experiments
Catalog No. 50-003-85727 Supplier Medchemexpress LLC Supplier No. HY17034B100MG
Medetomidine hydrochloride is an analytical standard of Medetomidine (hydrochloride), intended for research and analytical applications. This orally active α2-adrenoceptor agonist, with a Ki of 1.08 nM, exhibits sedative and analgesic effects, and can induce peripheral vasoconstriction. It serves as a reference standard for assays and is commonly utilized in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Analytical standard for research and analytical applications
  • Orally active α2-adrenoceptor agonist (Ki: 1.08 nM)
  • Provides sedative and analgesic effects
  • Induces peripheral vasoconstriction
  • Reference standard for assays
  • Suitable for HPLC, GC, and MS research experiments