Learn More
Medchemexpress LLC Medetomidine (hydrochloride) | 86347-15-1 | 99.8% | 236.74 | 25 MG
SDP
Catalog No. 5000385573
Encompass_Preferred
Medetomidine hydrochloride is an orally active α2-adrenoceptor agonist with a Ki of 1.08 nM. It exhibits sedative and analgesic effects and can cause peripheral vasoconstriction by activating α2 adrenoceptors on blood vessels.
  • Inhibits aldosterone release.
  • Activates a kicking response in Cyprids.
  • Increases cellular cAMP production.
  • Induces sedation in cats.
  • Shows sedative and analgesic effects in dogs.
  • Protects against Diazinon-induced toxicosis in mice.
Catalog No. 50-003-85573 Supplier Medchemexpress LLC Supplier No. 25MGHY17034B
Medetomidine hydrochloride is an orally active α2-adrenoceptor agonist with a Ki of 1.08 nM. It exhibits sedative and analgesic effects and can cause peripheral vasoconstriction by activating α2 adrenoceptors on blood vessels.
  • Inhibits aldosterone release.
  • Activates a kicking response in Cyprids.
  • Increases cellular cAMP production.
  • Induces sedation in cats.
  • Shows sedative and analgesic effects in dogs.
  • Protects against Diazinon-induced toxicosis in mice.