Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Medetomidine (hydrochloride) | 86347-15-1 | 99.8% | 236.74 | 5 MG
SDP

Catalog No. 5000384501
Encompass_Preferred

Medetomidine hydrochloride from MedChemExpress is provided for laboratory chemical applications and substance manufacturing. This product is a white to off-white solid.

  • Suitable for laboratory chemicals and substance manufacturing
  • White to off-white solid appearance
  • Store at 4°C, sealed and away from moisture
  • Purity of 99.8% confirmed by LCMS
  • For research use only; not validated for medical applications

Catalog No. 50-003-84501 Supplier Medchemexpress LLC Supplier No. HY17034B5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Medetomidine hydrochloride from MedChemExpress is provided for laboratory chemical applications and substance manufacturing. This product is a white to off-white solid.

  • Suitable for laboratory chemicals and substance manufacturing
  • White to off-white solid appearance
  • Store at 4°C, sealed and away from moisture
  • Purity of 99.8% confirmed by LCMS
  • For research use only; not validated for medical applications

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.