Learn More
Abcam Medetomidine hydrochloride, alpha2 adrenoceptor agonist, 50MG
Catalog No. 502707269
Encompass_Preferred
MW 236.74 Da, Purity >99%. Selective α₂ adrenoceptor agonist (Ki values for receptor subtypes are 3.89 (α₂A), 7.40 (α₂B), 12.3 (α₂C) and 0.76 nM (α₂D)). Crosses the blood-brain barrier. Potent sedative, hypnotic and analgesic properties in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7269 Supplier Abcam Supplier No. AB12078750MG
MW 236.74 Da, Purity >99%. Selective α₂ adrenoceptor agonist (Ki values for receptor subtypes are 3.89 (α₂A), 7.40 (α₂B), 12.3 (α₂C) and 0.76 nM (α₂D)). Crosses the blood-brain barrier. Potent sedative, hypnotic and analgesic properties in vivo.
The product is subject to the following: Abcam Restricted Use Statement