Learn More
BTNX Inc Medetomidine Test Strip - 100pk
Catalog No. NC3361242
Encompass_Preferred
The Rapid Response™ Medetomidine Test Strip (Liquid/Powder) is an easy to use, cost-effective dipstick test. It is a rapid, qualitative screening test that quickly and reliably detects medetomidine at cut-off levels above 1000 ng/mL. This drug test can provide accurate and fast results as soon as 1 minute. For forensic use only. Not an IVD.
Catalog No. NC3361242 Supplier BTNX Inc Supplier No. MED18S2100
The Rapid Response™ Medetomidine Test Strip (Liquid/Powder) is an easy to use, cost-effective dipstick test. It is a rapid, qualitative screening test that quickly and reliably detects medetomidine at cut-off levels above 1000 ng/mL. This drug test can provide accurate and fast results as soon as 1 minute. For forensic use only. Not an IVD.