Learn More
Cayman Chemical Medetomidine (hydrochloride), Purity: ≥98%, CAS: 86347-15-1, 50 mg
SDP
Catalog No. 502972494
Encompass_Preferred
An α2-AR agonist (Ki = 1.08 nM); selective for α2-ARs over α1-ARs (Ki = 1,750 nM); induces sedation in dogs
Catalog No. 50-297-2494 Supplier Cayman Chemical Supplier No. 1645450 MG
An α2-AR agonist (Ki = 1.08 nM); selective for α2-ARs over α1-ARs (Ki = 1,750 nM); induces sedation in dogs