Learn More
Medchemexpress LLC MEDI-570 10mg | 10mg
SDP
Catalog No. 5000324518
Encompass_Preferred
MEDI-570 is a humanized antibody expressed in CHO that targets ICOS/CD278 MEDI-570 contains huIgG1 heavy chain and hu light chain with a predicted molecular weight (MW) of 145 kDa The isotype control for MEDI-570 can be referenced as Human IgG1 kappa Isotype Control (HY-P99001)
Catalog No. 50-003-24518 Supplier Medchemexpress LLC Supplier No. HYP99062810MG
MEDI-570 is a humanized antibody expressed in CHO that targets ICOS/CD278 MEDI-570 contains huIgG1 heavy chain and hu light chain with a predicted molecular weight (MW) of 145 kDa The isotype control for MEDI-570 can be referenced as Human IgG1 kappa Isotype Control (HY-P99001)