Learn More
Medchemexpress LLC MEDI-570 5mg | 5mg
SDP
Catalog No. 5000325016
Encompass_Preferred
MEDI-570 is a humanized antibody expressed in CHO that targets ICOS/CD278 MEDI-570 contains huIgG1 heavy chain and hu light chain with a predicted molecular weight (MW) of 145 kDa The isotype control for MEDI-570 can be referenced as Human IgG1 kappa Isotype Control (HY-P99001)
Catalog No. 50-003-25016 Supplier Medchemexpress LLC Supplier No. HYP9906285MG
MEDI-570 is a humanized antibody expressed in CHO that targets ICOS/CD278 MEDI-570 contains huIgG1 heavy chain and hu light chain with a predicted molecular weight (MW) of 145 kDa The isotype control for MEDI-570 can be referenced as Human IgG1 kappa Isotype Control (HY-P99001)