Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Medi-First™ 10 Person Plastic ANSI-A First Aid Kit

Catalog No. 19033691 Shop All Fisherbrand Products
Change view
Click to view available options
Material:
Plastic
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-033-691 Plastic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-033-691
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

This compact kit provides inexpensive first aid for up to 10 people, and can be wall mounted or portable with an easy-to-hold carrying handle. Meets ANSI Z308.1-2021 Class A.

  • Kit fills subject to change
  • Meets ANSI Z308.1-2021
  • Kit contains:
    • 1 Adhesive Bandage (1 x 3 in.)
    • 16 Plastic Strips
    • 2 Adhesive Tapes, (1/2 in. x 2.5 yd.)
    • 3 Ammonia Wipes
    • 10 Antiseptic Wipes
    • 10 Burn Cream with Lidocaine
    • 1 Burn Dressing (4 x 4 in.)
    • 1 Small Cold Pack
    • 2 Combine ABD Pads (5 x 9 in.)
    • 1 CPR Mask/Breathing Barrier
    • 2 Eye Pads
    • 1 oz. Eye Wash
    • 1 First Aid Handbook
    • 2 Sterile Gauze Pads (3 x 3 in.)
    • 1 Gauze Roll (2 in. x 5 yd.)
    • 10 Hand Sanitizer
    • 3 Insect Sting Relief Wipes
    • 2 Disposable, Powder-free Vinyl Glove Pairs, Size: Large
    • 1 Silver Rescue Blanket (52 x 84 in.)
    • 1 Non-sterile Triangular Bandage with Pins
    • 10 Triple Antibiotic Ointment
    • 1 Disposable Tweezers
    • 1 Wire Scissors 4.5 in.

Specifications

Product Type First Aid Kit
For Use With (Application) First Aid Response, OSHA Compliance
Certifications/Compliance ANSI Z308.1-2021
Color White
Material Plastic
Sterility Non-sterile
Mounting Wall Mountable
Dimensions (L x W x H) 9.19 x 6.25 x 2.75 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.