Learn More
CTL Scientific Supply Corp MEDI-TEST COMBI 10 VET
SDP
Catalog No. 50300394 Shop All CTL Scientific Supply Products
Encompass_Preferred
Urine test strip for veterinary applications including glucose, keytones, protein, blood, leukocytes, nitrite, specific gravity, pH, bilirubin and urobilinogen. 100 tests/box
Catalog No. 50-300-394 Supplier CTL Scientific Supply Corp Supplier No. 930870
Urine test strip for veterinary applications including glucose, keytones, protein, blood, leukocytes, nitrite, specific gravity, pH, bilirubin and urobilinogen. 100 tests/box