Learn More
Bellco Glass, Inc. Media Addition Bottle,2L
SDP
Catalog No. 501944875
Encompass_Preferred
The Media Addition Bottle is designed for holding media and comes with Stainless Steel 2-port assembly for attachment of tubing for connection to a pump to add media to a bioreactor vessel or spinner flask. MADE IN THE USA. Specifications:Size: 2L316L Stainless Steel Port AssemblyRa 32 or Better FinishType 1A Borosilicate Glass BottleCase Quantity: 1
Catalog No. 50-194-4875 Supplier Bellco Glass, Inc. Supplier No. 196483200
The Media Addition Bottle is designed for holding media and comes with Stainless Steel 2-port assembly for attachment of tubing for connection to a pump to add media to a bioreactor vessel or spinner flask. MADE IN THE USA. Specifications:Size: 2L316L Stainless Steel Port AssemblyRa 32 or Better FinishType 1A Borosilicate Glass BottleCase Quantity: 1