Learn More
Chemglass Life Sciences Adapter, Addition Pipe, M10, for 1-15L Bio Reactors
SDP
Catalog No. 501294430
Encompass
Adapter, Addition Pipe, M10, for 2-7L Bio Reactors. This adapter can be used to add fluids or gases to the bio reactor. The portion of the adapter that is within the reactor has a short drip tip tube that can accept tubing to reach further into the listed capacity reactor.Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors.Applikon is a registered trademark of Applikon Biotechnology, Inc. Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.
Catalog No. 50-129-4430 Supplier Chemglass Life Sciences Supplier No. CLS138031
Adapter, Addition Pipe, M10, for 2-7L Bio Reactors. This adapter can be used to add fluids or gases to the bio reactor. The portion of the adapter that is within the reactor has a short drip tip tube that can accept tubing to reach further into the listed capacity reactor.Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors.Applikon is a registered trademark of Applikon Biotechnology, Inc. Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.