Learn More
FUJIFILM Biosciences BalanCD CHO Growth A Medium - Powder
Catalog No. NC1786903
Encompass
BalanCD CHO Growth A is a chemically-defined, animal component-free growth medium designed to increase process yields of antibodies and recombinant proteins in Chinese Hamster Ovary (CHO) cells. This formulation was developed using FUJIFILM Irvine Scientific’s Rational Culture Media Design strategy to achieve enhanced performance of growth and production in batch cultures. This medium is designed to be used in conjunction with BalanCD CHO Feed 4 for optimal performance in fed-batch cultures.
Catalog No. NC1786903 Supplier FUJIFILM Biosciences Supplier No. 94120100L
Add to Cart
Add to Cart
Request a Quote
BalanCD CHO Growth A is a chemically-defined, animal component-free growth medium designed to increase process yields of antibodies and recombinant proteins in Chinese Hamster Ovary (CHO) cells. This formulation was developed using FUJIFILM Irvine Scientific’s Rational Culture Media Design strategy to achieve enhanced performance of growth and production in batch cultures. This medium is designed to be used in conjunction with BalanCD CHO Feed 4 for optimal performance in fed-batch cultures.