Learn More
Alkali Scientific CellPro™ Square Media Bottles, 1000mL, Sterile, 12/Tray, 24/cs
SDP
Catalog No. 502335908
Encompass_Preferred
CellPro™ square media bottles, composed of high-strength plastic material, are intended for storing and transporting general reagents and cell culture reagents. The square-shaped bottles are designed to maximize space saving, durability, strength, and clarity as close to glass as possible. They have molded-in graduation scales and are non-pyrogenic, non-cytotoxic, DNase/RNase-free, and human DNA-free.
Catalog No. 50-233-5908 Supplier Alkali Scientific Supplier No. VB1000
CellPro™ square media bottles, composed of high-strength plastic material, are intended for storing and transporting general reagents and cell culture reagents. The square-shaped bottles are designed to maximize space saving, durability, strength, and clarity as close to glass as possible. They have molded-in graduation scales and are non-pyrogenic, non-cytotoxic, DNase/RNase-free, and human DNA-free.