Learn More
Grainger Media Bottle 111 mm H 114.3 mm Dia PK5
Catalog No. 502750410
Encompass_Preferred
The Benchmark Scientific Media Bottle is made of borosilicate glass and has a capacity of 250mL. It has a wide mouth opening and polypropylene screw cap closure. The bottles are designed for general purpose use in the laboratory and can withstand temperatures up to 500°F. They are autoclavable and have volumetric graduations up to 250mL. The dimensions are 2-7/8" diameter x 5-1/2" height. These media bottles provide an economical way to store powders, liquids and other laboratory media. The bottles are sold in cases of 144.
Catalog No. 50-275-0410 Supplier Grainger Supplier No. 14N164
The Benchmark Scientific Media Bottle is made of borosilicate glass and has a capacity of 250mL. It has a wide mouth opening and polypropylene screw cap closure. The bottles are designed for general purpose use in the laboratory and can withstand temperatures up to 500°F. They are autoclavable and have volumetric graduations up to 250mL. The dimensions are 2-7/8" diameter x 5-1/2" height. These media bottles provide an economical way to store powders, liquids and other laboratory media. The bottles are sold in cases of 144.