Learn More
Grainger Media Bottle 171.45mm H 82.55mm Dia PK10
Catalog No. 502749416
Encompass_Preferred
The Benchmark Scientific Media Bottle is made of borosilicate glass and has a capacity of 250mL. It has a wide mouth opening and polypropylene screw cap closure. The bottles are designed for general purpose use in laboratories and can withstand temperatures up to 500°F. They are autoclavable and have volumetric graduations up to 250mL. The dimensions are 3" diameter x 5.7" height. These media bottles provide an economical option for storing powders, liquids, cultures, and more. The bottles are sold in cases of 144.
Catalog No. 50-274-9416 Supplier Grainger Supplier No. 14N162
The Benchmark Scientific Media Bottle is made of borosilicate glass and has a capacity of 250mL. It has a wide mouth opening and polypropylene screw cap closure. The bottles are designed for general purpose use in laboratories and can withstand temperatures up to 500°F. They are autoclavable and have volumetric graduations up to 250mL. The dimensions are 3" diameter x 5.7" height. These media bottles provide an economical option for storing powders, liquids, cultures, and more. The bottles are sold in cases of 144.