Learn More
Grainger Media Bottle 215.9mm H 95.25mm Dia PK10
Catalog No. 502750409
Encompass_Preferred
The Benchmark Scientific Media Bottle is made of borosilicate glass and has a capacity of 250mL. It has a wide mouth opening and polypropylene screw cap closure. The bottles are designed for general purpose use in laboratories and can withstand temperatures up to 500°F. They are autoclavable and have volumetric graduations. The dimensions are 2-7/8" diameter x 5-1/8" height. These media bottles provide an economical way to store powders, liquids and other laboratory media.
Catalog No. 50-275-0409 Supplier Grainger Supplier No. 14N163
The Benchmark Scientific Media Bottle is made of borosilicate glass and has a capacity of 250mL. It has a wide mouth opening and polypropylene screw cap closure. The bottles are designed for general purpose use in laboratories and can withstand temperatures up to 500°F. They are autoclavable and have volumetric graduations. The dimensions are 2-7/8" diameter x 5-1/8" height. These media bottles provide an economical way to store powders, liquids and other laboratory media.