Learn More
Grainger Media Bottle 4 mm H 2.2 mm Dia PK10
Catalog No. 502749628
Encompass_Preferred
The Grainger 14N160 is a 14 inch diameter, 160 grit aluminum oxide cutoff wheel that is rated for 10,000 rpm max. It has a 1/4 inch arbor hole and is 1/16 inch thick. The wheel is designed for general purpose cutting of mild steel, stainless steel, aluminum and other metals. It should be used with a guard for safety. The wheel allows for fast, smooth cutting action.
Catalog No. 50-274-9628 Supplier Grainger Supplier No. 14N160
The Grainger 14N160 is a 14 inch diameter, 160 grit aluminum oxide cutoff wheel that is rated for 10,000 rpm max. It has a 1/4 inch arbor hole and is 1/16 inch thick. The wheel is designed for general purpose cutting of mild steel, stainless steel, aluminum and other metals. It should be used with a guard for safety. The wheel allows for fast, smooth cutting action.