Learn More
Bellco Glass, Inc. Media Bottle Only,15L No Cap or Ring
SDP
Catalog No. 501945800
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. These media storage bottles are chemically resistant ans stable. They are suitable for storage as well as for mixing and sampling. They are autoclavable and have white graduations and marking spot. DIMENSIONS: 268mm X 450mm TALL
Catalog No. 50-194-5800 Supplier Bellco Glass, Inc. Supplier No. 563768000
Manufactured from Type 1 Class A Borosilicate Glass. These media storage bottles are chemically resistant ans stable. They are suitable for storage as well as for mixing and sampling. They are autoclavable and have white graduations and marking spot. DIMENSIONS: 268mm X 450mm TALL