Learn More
Bellco Glass, Inc. Media Bottle Only,2000mL No Cap or Ring
SDP
Catalog No. 501945797
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. These media storage bottles are chemically resistant ans stable. They are suitable for storage as well as for mixing and sampling. They are autoclavable and have white graduations and marking spot. DIMENSIONS: 136mm X 260mm TALL. MADE IN THE USA
Catalog No. 50-194-5797 Supplier Bellco Glass, Inc. Supplier No. 563762000
Manufactured from Type 1 Class A Borosilicate Glass. These media storage bottles are chemically resistant ans stable. They are suitable for storage as well as for mixing and sampling. They are autoclavable and have white graduations and marking spot. DIMENSIONS: 136mm X 260mm TALL. MADE IN THE USA