Learn More
Bellco Glass, Inc. Media Bottle Only,500mL No Cap or Ring
SDP
Catalog No. 501945795
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. These media storage bottles are chemically resistant ans stable. They are suitable for storage as well as for mixing and sampling. They are autoclavable and have white graduations and marking spot. DIMENSIONS: 86mm X 176mm TALL MADE IN THE USA
Catalog No. 50-194-5795 Supplier Bellco Glass, Inc. Supplier No. 563760500
Manufactured from Type 1 Class A Borosilicate Glass. These media storage bottles are chemically resistant ans stable. They are suitable for storage as well as for mixing and sampling. They are autoclavable and have white graduations and marking spot. DIMENSIONS: 86mm X 176mm TALL MADE IN THE USA